This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03422846_m1
See other UBE3A GE Assays ›
Gene Symbol:
ube3a
Gene Name:
ubiquitin protein ligase E3A
Gene Aliases:
im:7140733, zgc:92173
Chromosome Location:
Chr.6: 38773406 - 38790064 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03422846_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ube3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007318.1 NP_001007319.1 3 - 4 514 83
GenBank mRNA BC085646.1 - 3 - 4 514 83
BC155156.1 - 3 - 4 307 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
transferase activity
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0