This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03423568_m1
See other NAA20 GE Assays ›
Gene Symbol:
naa20
Gene Name:
N-alpha-acetyltransferase 20, NatB catalytic subunit
Gene Aliases:
im:7150897, nat5, zgc:110819
Chromosome Location:
Chr.20: 49105429 - 49117401 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03423568_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: naa20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014329.2 NP_001014351.1 5 - 6 476 69
GenBank mRNA BC091957.1 - 5 - 6 480 69
BC129286.1 - 5 - 6 486 69
BC164930.1 - 5 - 6 482 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

N-terminal peptidyl-methionine acetylation
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0