This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03426822_m1
See other GMPR2 GE Assays ›
Gene Symbol:
gmpr2
Gene Name:
guanosine monophosphate reductase 2
Gene Aliases:
wu:fb63f02, zgc:136869
Chromosome Location:
Chr.7: 24197457 - 24204655 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03426822_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gmpr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040304.1 NP_001035394.1 3 - 4 294 75
GenBank mRNA AI497282.1 - 3 - 4 291 75
BC115299.1 - 3 - 4 294 75
CK674610.1 - 3 - 4 308 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase metabolic process
purine nucleotide metabolic process
nucleotide metabolic process
oxidation-reduction process
catalytic activity
GMP reductase activity
oxidoreductase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline