This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03427923_m1
See other CISH GE Assays ›
Gene Symbol:
cish
Gene Name:
cytokine inducible SH2-containing protein
Gene Aliases:
cis, cish.a, ptk9l, twf1l, twf2a, wu:fk57g04, zgc:154062, zgc:91817
Chromosome Location:
Chr.6: 41503760 - 41506753 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03427923_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cish
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076617.2 NP_001070085.1 2 - 3 402 123
XM_005166065.3 XP_005166122.1 2 - 3 747 123
GenBank mRNA AM040611.1 - 2 - 3 146 123
BC124528.1 - 2 - 3 411 123
EF195760.1 - 2 - 3 402 123
FR749998.1 - 2 - 3 439 123

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
intracellular signal transduction
negative regulation of lymphocyte differentiation
negative regulation of myeloid cell differentiation
negative regulation of erythrocyte differentiation
negative regulation of JAK-STAT cascade
phosphatidylinositol phosphorylation
negative regulation of hemopoiesis
1-phosphatidylinositol-3-kinase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline