This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03430949_m1
See other RAC3B GE Assays ›
Gene Symbol:
rac3b
Gene Name:
Rac family small GTPase 3b
Gene Aliases:
zgc:175209
Chromosome Location:
Chr.12: 3022872 - 3042029 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03430949_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rac3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320406.1 NP_001307335.1 1 - 2 151 70
GenBank mRNA BC155847.1 - 1 - 2 141 70
EH436479.1 - 1 - 2 181 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC155847.1, NM_001320406.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
establishment or maintenance of cell polarity
small GTPase mediated signal transduction
regulation of cell shape
rhombomere boundary formation
actin cytoskeleton organization
cortical cytoskeleton organization
actomyosin structure organization
regulation of actin cytoskeleton organization
nucleotide binding
GTPase activity
GTP binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline