This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03431454_m1
See other TNFRSF18 GE Assays ›
Gene Symbol:
tnfrsf18
Gene Name:
tumor necrosis factor receptor superfamily, member 18
Gene Aliases:
-
Chromosome Location:
Chr.6: 42975279 - 42984693 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03431454_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tnfrsf18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113524.1 NP_001106996.1 5 - 6 733 95
XM_005166063.4 XP_005166120.1 5 - 6 1589 95
GenBank mRNA EU099311.1 - 5 - 6 733 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of apoptotic process
positive regulation of cell adhesion
tumor necrosis factor-activated receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0