This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03433120_m1
See other OPRM1 GE Assays ›
Gene Symbol:
oprm1
Gene Name:
opioid receptor, mu 1
Gene Aliases:
ZFOR2, or2
Chromosome Location:
Chr.13: 23843614 - 23882977 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03433120_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: oprm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131707.2 NP_571782.2 1 - 2 374 71
GenBank mRNA AF132813.1 - 1 - 2 283 71
BC163729.1 - 1 - 2 374 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
sensory perception of pain
opioid receptor signaling pathway
G-protein alpha-subunit binding
G-protein coupled receptor activity
beta-endorphin receptor activity
opioid receptor activity
G-protein beta-subunit binding
enkephalin receptor activity
morphine receptor activity
peptide binding
neuropeptide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0