This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434614_m1
See other GMNN GE Assays ›
Gene Symbol:
gmnn
Gene Name:
geminin DNA replication inhibitor
Gene Aliases:
sb:cb254, zgc:66217
Chromosome Location:
Chr.19: 538701 - 543908 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434614_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gmnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200086.1 NP_956380.1 5 - 6 457 69
XM_021468008.1 XP_021323683.1 4 - 5 377 69
GenBank mRNA BC049069.1 - 5 - 6 455 69
BC055552.1 - 5 - 6 457 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
negative regulation of DNA replication
determination of pancreatic left/right asymmetry
negative regulation of cell cycle
cilium morphogenesis
determination of heart left/right asymmetry
Kupffer's vesicle development
determination of liver left/right asymmetry
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0