This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434642_m1
See other EPB41L5 GE Assays ›
Gene Symbol:
epb41l5
Gene Name:
erythrocyte membrane protein band 4.1 like 5
Gene Aliases:
dZ104F18.2, dZ104F18.3, si:dz104f18.2, si:dz104f18.3, si:dz37o5.2
Chromosome Location:
Chr.9: 154837 - 217168 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434642_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: epb41l5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200089.2 NP_956383.1 7 - 8 666 61
GenBank mRNA AY423009.1 - 7 - 8 679 61
AY590884.1 - 6 - 7 506 61
BC066560.1 - 7 - 8 666 61
BC155177.1 - 7 - 8 655 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

kidney development
dorsal motor nucleus of vagus nerve development
actomyosin structure organization
cellular protein complex localization
establishment or maintenance of apical/basal cell polarity
establishment or maintenance of epithelial cell apical/basal polarity
maintenance of epithelial cell apical/basal polarity
positive regulation of Notch signaling pathway
embryonic eye morphogenesis
embryonic retina morphogenesis in camera-type eye
cytoskeletal protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline