This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03434757_m1
See other PEMT GE Assays ›
Gene Symbol:
pemt
Gene Name:
phosphatidylethanolamine N-methyltransferase
Gene Aliases:
zgc:55479
Chromosome Location:
Chr.12: 1286531 - 1388444 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03434757_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pemt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200158.1 NP_956452.1 3 - 4 556 69
GenBank mRNA BC045355.1 - 3 - 4 556 69
BC165059.1 - 479 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylcholine biosynthetic process
phospholipid biosynthetic process
methylation
phosphatidyl-N-methylethanolamine N-methyltransferase activity
phosphatidylethanolamine N-methyltransferase activity
methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
transferase activity
phosphatidyl-N-dimethylethanolamine N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0