This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03435817_m1
See other SOCS2 GE Assays ›
Gene Symbol:
socs2
Gene Name:
suppressor of cytokine signaling 2
Gene Aliases:
si:dkey-202b22.1
Chromosome Location:
Chr.4: 18431531 - 18436958 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03435817_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: socs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114550.1 NP_001108022.1 1 - 2 124 54
XM_005164748.4 XP_005164805.1 2 - 3 701 54
GenBank mRNA EF195761.1 - 1 - 2 124 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
protein ubiquitination
intracellular signal transduction
negative regulation of JAK-STAT cascade
phosphatidylinositol phosphorylation
insulin-like growth factor receptor binding
JAK pathway signal transduction adaptor activity
1-phosphatidylinositol-3-kinase regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline