This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04951574_m1
See other AIMP1 GE Assays ›
Gene Symbol:
AIMP1
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 119077529 - 119173847 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04951574_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001309207.1 NP_001296136.1 3 - 4 408 114
XM_023627008.1 XP_023482776.1 547 114
GenBank mRNA JL615910.1 - 4 - 5 446 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Equus caballus Probe spans exons

Panther Classification:

Molecular Function -

translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline