This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec04952672_m1
See other SLCO1B3 GE Assays ›
Gene Symbol:
SLCO1B3
Gene Name:
solute carrier organic anion transporter family member 1B3
Gene Aliases:
Oatp1b4
Chromosome Location:
Chr.6: 48670761 - 48748040 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec04952672_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1B3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317251.1 NP_001304180.1 13 - 14 1893 85
XM_005610968.3 XP_005611025.1 2322 85
XM_023642200.1 XP_023497968.1 2327 85
XM_023642201.1 XP_023497969.1 2184 85
GenBank mRNA LN833581.1 - 13 - 14 1893 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Equus caballus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0