This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06952034_m1
See other LOC100058358 GE Assays ›
Gene Symbol:
LOC100058358
Gene Name:
actin-related protein 2/3 complex subunit 4
Gene Aliases:
ARPC4, TTLL3
Chromosome Location:
Chr.16: 8463587 - 8497254 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06952034_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100058358
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023620002.1 XP_023475770.1 1202 91
XM_023620003.1 XP_023475771.1 1199 91
XM_023620004.1 XP_023475772.1 1204 91
XM_023620005.1 XP_023475773.1 1204 91
XM_023620006.1 XP_023475774.1 1203 91
XM_023620007.1 XP_023475775.1 1203 91
XM_023620008.1 XP_023475776.1 1204 91
XM_023620009.1 XP_023475777.1 846 91
XM_023620010.1 XP_023475778.1 750 91
XM_023620011.1 XP_023475779.1 485 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline