This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06952036_m1
See other LOC100058358 GE Assays ›
Gene Symbol:
LOC100058358
Gene Name:
actin-related protein 2/3 complex subunit 4
Gene Aliases:
ARPC4, TTLL3
Chromosome Location:
Chr.16: 8463587 - 8497254 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06952036_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100058358
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023620002.1 XP_023475770.1 1543 57
XM_023620003.1 XP_023475771.1 1540 57
XM_023620004.1 XP_023475772.1 1545 57
XM_023620005.1 XP_023475773.1 1545 57
XM_023620006.1 XP_023475774.1 1544 57
XM_023620007.1 XP_023475775.1 1544 57
XM_023620008.1 XP_023475776.1 1545 57
XM_023620009.1 XP_023475777.1 1187 57
XM_023620010.1 XP_023475778.1 1091 57
XM_023620011.1 XP_023475779.1 826 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline