This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06961437_m1
See other TRAT1 GE Assays ›
Gene Symbol:
TRAT1
Gene Name:
T cell receptor associated transmembrane adaptor 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 50860557 - 50895714 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06961437_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005602016.3 XP_005602073.1 176 150
GenBank mRNA JL634090.1 - 1 - 2 196 150

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005602016.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Equus caballus Probe spans exons

Panther Classification:

Molecular Function -

defense/immunity protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline