This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06981122_g1
See other HDHD3 GE Assays ›
Gene Symbol:
HDHD3
Gene Name:
haloacid dehalogenase like hydrolase domain containing 3
Gene Aliases:
-
Chromosome Location:
Chr.25: 18696846 - 18699696 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06981122_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005605714.3 XP_005605771.1 319 89
XM_014735977.2 XP_014591463.1 410 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005605714.3, XM_014735977.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline