This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06998034_m1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
-
Chromosome Location:
Chr.7: 23390392 - 23417601 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06998034_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005611582.3 XP_005611639.1 529 100
XM_005611586.3 XP_005611643.1 529 100
XM_014741370.2 XP_014596856.1 517 100
XM_023644853.1 XP_023500621.1 298 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005611582.3, XM_005611586.3, XM_014741370.2, XM_023644853.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0