This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec06998037_m1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
-
Chromosome Location:
Chr.7: 23390392 - 23417601 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec06998037_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005611582.3 XP_005611639.1 1429 100
XM_005611587.3 XP_005611644.1 317 100
XM_014741370.2 XP_014596856.1 1417 100
XM_014741371.2 XP_014596857.1 1355 100
XM_014741372.2 XP_014596858.1 162 100
XM_023644853.1 XP_023500621.1 1198 100
GenBank mRNA JL640141.1 - 2 - 3 1083 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014741372.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Equus caballus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0