Assay Details
| Gene Symbol: P2RY13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489171.5 | XP_001489221.1 | 127 | 77 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2RY13| Gene Symbol: P2RY13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489171.5 | XP_001489221.1 | 127 | 77 | ||
Set Membership: |
|