This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07038529_m1
See other PCED1B GE Assays ›
Gene Symbol:
PCED1B
Gene Name:
PC-esterase domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.6: 65858726 - 65962146 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07038529_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCED1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005611103.2 XP_005611160.2 419 87
XM_014740964.2 XP_014596450.2 2991 87
XM_014740966.2 XP_014596452.2 323 87
XM_023643435.1 XP_023499203.1 413 87
XM_023643436.1 XP_023499204.1 342 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005611103.2, XM_014740964.2, XM_014740966.2, XM_023643435.1, XM_023643436.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0