This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07041005_m1
See other LIPJ GE Assays ›
Gene Symbol:
LIPJ
Gene Name:
lipase family member J
Gene Aliases:
-
Chromosome Location:
Chr.1: 40275673 - 40315166 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07041005_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPJ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001501539.4 XP_001501589.3 1705 107
XM_023643145.1 XP_023498913.1 1831 107
XM_023643148.1 XP_023498916.1 1267 107
XM_023643149.1 XP_023498917.1 1930 107
XM_023643160.1 XP_023498928.1 1685 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Equus caballus

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0