This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07041958_m1
See other ZNF239 GE Assays ›
Gene Symbol:
ZNF239
Gene Name:
zinc finger protein 239
Gene Aliases:
-
Chromosome Location:
Chr.1: 71089584 - 71123138 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07041958_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014733872.2 XP_014589358.2 198 94
XM_014733874.2 XP_014589360.2 198 94
XM_014733875.2 XP_014589361.2 198 94
XM_014733876.2 XP_014589362.2 198 94
XM_014733877.2 XP_014589363.2 269 94
XM_014733879.2 XP_014589365.2 198 94
XM_023646521.1 XP_023502289.1 198 94
XM_023646542.1 XP_023502310.1 133 94
XM_023646550.1 XP_023502318.1 239 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014733872.2, XM_014733874.2, XM_014733875.2, XM_014733876.2, XM_014733877.2, XM_014733879.2, XM_023646521.1, XM_023646542.1, XM_023646550.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Equus caballus

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0