This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07042566_m1
See other PTPN20 GE Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase non-receptor type 20
Gene Aliases:
PTPN20B
Chromosome Location:
Chr.1: 83335624 - 83448468 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07042566_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005602729.3 XP_005602786.1 761 86
XM_005602732.3 XP_005602789.1 761 86
XM_014734101.2 XP_014589587.1 761 86
XM_023648426.1 XP_023504194.1 762 86
XM_023648429.1 XP_023504197.1 765 86
XM_023648433.1 XP_023504201.1 770 86
XM_023648436.1 XP_023504204.1 745 86
XM_023648440.1 XP_023504208.1 752 86
XM_023648447.1 XP_023504215.1 671 86
XM_023648449.1 XP_023504217.1 682 86
XM_023648455.1 XP_023504223.1 676 86
XM_023648464.1 XP_023504232.1 762 86
XM_023648478.1 XP_023504246.1 761 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Equus caballus

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0