Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014741939.2 | XP_014597425.1 | 328 | 104 | ||
| XM_023649864.1 | XP_023505632.1 | 363 | 104 | |||
| XM_023649865.1 | XP_023505633.1 | 363 | 104 | |||
| XM_023649866.1 | XP_023505634.1 | 363 | 104 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014741939.2 transcripts.
Set Membership: |
|

ZNF180
Genomic Map