This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07050211_m1
See other ZNF615 GE Assays ›
Gene Symbol:
ZNF615
Gene Name:
zinc finger protein 615
Gene Aliases:
ZNF350
Chromosome Location:
Chr.10: 21995265 - 22066584 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07050211_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF615
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014730240.2 XP_014585726.2 450 94
XM_014730249.2 XP_014585735.2 171 94
XM_014730256.2 XP_014585742.2 498 94
XM_014730264.2 XP_014585750.2 456 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014730240.2, XM_014730249.2, XM_014730256.2, XM_014730264.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Equus caballus

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline