This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07050258_g1
See other OSCAR GE Assays ›
Gene Symbol:
OSCAR
Gene Name:
osteoclast associated Ig-like receptor
Gene Aliases:
-
Chromosome Location:
Chr.10: 23581302 - 23586898 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07050258_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OSCAR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023650350.1 XP_023506118.1 131 81
XM_023650351.1 XP_023506119.1 134 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_023650350.1, XM_023650351.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Equus caballus

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0