This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07050553_m1
See other ZNF544 GE Assays ›
Gene Symbol:
ZNF544
Gene Name:
zinc finger protein 544
Gene Aliases:
-
Chromosome Location:
Chr.10: 28320392 - 28340653 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07050553_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF544
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023650621.1 XP_023506389.1 1577 108
XM_023650622.1 XP_023506390.1 1538 108
XM_023650624.1 XP_023506392.1 1688 108
XM_023650625.1 XP_023506393.1 1649 108
XM_023650626.1 XP_023506394.1 1471 108
XM_023650627.1 XP_023506395.1 775 108
XM_023650628.1 XP_023506396.1 807 108
XM_023650629.1 XP_023506397.1 507 108
XM_023650630.1 XP_023506398.1 1577 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0