This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07050554_m1
See other ZNF544 GE Assays ›
Gene Symbol:
ZNF544
Gene Name:
zinc finger protein 544
Gene Aliases:
-
Chromosome Location:
Chr.10: 28320392 - 28340653 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07050554_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF544
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_023650621.1 XP_023506389.1 1666 65
XM_023650622.1 XP_023506390.1 1627 65
XM_023650624.1 XP_023506392.1 1777 65
XM_023650625.1 XP_023506393.1 1738 65
XM_023650626.1 XP_023506394.1 1560 65
XM_023650627.1 XP_023506395.1 864 65
XM_023650628.1 XP_023506396.1 896 65
XM_023650629.1 XP_023506397.1 596 65
XM_023650630.1 XP_023506398.1 1666 65
XM_023650631.1 XP_023506399.1 468 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_023650631.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Equus caballus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0