This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ec07061505_m1
See other BATF2 GE Assays ›
Gene Symbol:
BATF2
Gene Name:
basic leucine zipper ATF-like transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 28804392 - 28812696 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID Ec07061505_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001490074.4 XP_001490124.1 2221 96
XM_005598293.3 XP_005598350.1 2227 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001490074.4, XM_005598293.3 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Equus caballus

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline