This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03311112_m1
See other ATP5H GE Assays ›
Gene Symbol:
ATP5H
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit D
Gene Aliases:
ATP5PD
Chromosome Location:
Chr.18: 10624366 - 10627074 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03311112_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001232598.4 XP_001232599.1 4 - 5 346 86
XM_001232613.3 XP_001232614.1 4 - 5 393 86
GenBank mRNA BX930755.1 - 3 - 4 471 86
BX932142.2 - 3 - 4 532 86
BX934512.1 - 4 - 5 372 86
CR338739.1 - 3 - 4 309 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0