This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03311395_m1
See other IAH1 GE Assays ›
Gene Symbol:
IAH1
Gene Name:
isoamyl acetate-hydrolyzing esterase 1 homolog
Gene Aliases:
-
Chromosome Location:
Chr.3: 96209795 - 96213527 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03311395_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IAH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277729.1 NP_001264658.1 5 - 6 557 62
XM_025148779.1 XP_025004547.1 5 - 6 700 62
XM_025148780.1 XP_025004548.1 4 - 5 444 62
GenBank mRNA BX930468.1 - 5 - 6 557 62
BX931783.1 - 5 - 6 571 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

esterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0