This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03311676_m1
See other SSR3 GE Assays ›
Gene Symbol:
SSR3
Gene Name:
signal sequence receptor subunit 3
Gene Aliases:
-
Chromosome Location:
Chr.9: 23034760 - 23038625 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03311676_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277475.1 NP_001264404.1 4 - 5 548 85
GenBank mRNA BU322139.1 - 4 - 5 548 85
BX932955.1 - 4 - 5 496 85
BX933619.1 - 4 - 5 496 85
BX933663.1 - 4 - 5 543 85
BX934097.1 - 4 - 5 528 85
BX935222.1 - 4 - 5 503 85
BX935245.1 - 4 - 5 519 85
BX935248.2 - 4 - 5 528 85
BX935252.1 - 4 - 5 506 85
BX935261.1 - 4 - 5 543 85
BX935333.1 - 4 - 5 522 85
BX935792.1 - 4 - 5 544 85
BX936220.1 - 4 - 5 506 85
BX936255.1 - 4 - 5 522 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0