This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03312376_m1
See other ECI2 GE Assays ›
Gene Symbol:
ECI2
Gene Name:
enoyl-CoA delta isomerase 2
Gene Aliases:
PECI
Chromosome Location:
Chr.2: 65891511 - 65921167 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03312376_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001231828.5 XP_001231829.1 2 - 3 273 71
XM_004939668.3 XP_004939725.1 3 - 4 310 71
XM_015276009.2 XP_015131495.1 4 - 5 373 71
XM_025147668.1 XP_025003436.1 4 - 5 516 71
XM_025147669.1 XP_025003437.1 4 - 5 1275 71
XM_025147670.1 XP_025003438.1 5 - 6 609 71
XM_025147671.1 XP_025003439.1 5 - 6 1511 71
XM_025147672.1 XP_025003440.1 3 - 4 476 71
GenBank mRNA BX935815.1 - 2 - 3 224 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline