This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03312548_m1
See other PNLIPRP1 GE Assays ›
Gene Symbol:
PNLIPRP1
Gene Name:
pancreatic lipase related protein 1
Gene Aliases:
PNLIP
Chromosome Location:
Chr.6: 29804113 - 29815115 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03312548_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PNLIPRP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277382.1 NP_001264311.1 11 - 12 1194 62
XM_015288675.2 XP_015144161.1 10 - 11 1270 62
GenBank mRNA BX931579.1 - 10 - 11 1177 62
BX931581.1 - 11 - 12 1197 62
BX931592.1 - 9 - 10 950 62
BX931609.1 - 11 - 12 1196 62
BX933626.2 - 10 - 11 1172 62
BX950424.1 - 10 - 11 1176 62
DQ334850.1 - 9 - 10 1120 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0