This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03312987_m1
See other MTRF1 GE Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translation release factor 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 167195780 - 167214311 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03312987_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTRF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004938663.2 XP_004938720.1 7 - 8 1057 60
XM_004938664.3 XP_004938721.1 7 - 8 1101 60
XM_004938665.3 XP_004938722.1 7 - 8 1295 60
XM_004938666.2 XP_004938723.2 7 - 8 1057 60
XM_004938667.2 XP_004938724.2 6 - 7 949 60
XM_015276791.2 XP_015132277.1 8 - 9 1399 60
XM_015276793.2 XP_015132279.1 7 - 8 1137 60
XM_025146786.1 XP_025002554.1 7 - 8 1050 60
XM_025146787.1 XP_025002555.1 7 - 8 1101 60
XM_025146788.1 XP_025002556.1 7 - 8 1105 60
XM_417026.6 XP_417026.3 7 - 8 1058 60
GenBank mRNA BX929495.2 - 6 - 7 942 60
BX930267.2 - 4 - 5 795 60
CR523082.1 - 3 - 4 1098 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

translation release factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0