This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03313124_m1
See other HMGCL GE Assays ›
Gene Symbol:
HMGCL
Gene Name:
3-hydroxy-3-methylglutaryl-CoA lyase
Gene Aliases:
HL
Chromosome Location:
Chr.23: 5743508 - 5747131 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03313124_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMGCL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198714.1 NP_001185643.1 3 - 4 261 66
NM_001198715.1 NP_001185644.1 3 - 4 270 66
NM_001198716.1 NP_001185645.1 3 - 4 261 66
NM_001198717.1 NP_001185646.1 3 - 4 195 66
XM_025142963.1 XP_024998731.1 2 - 3 267 66
XM_025142964.1 XP_024998732.1 3 - 4 258 66
GenBank mRNA BX929535.1 - 3 - 4 243 66
BX931055.2 - 3 - 4 241 66
BX931402.1 - 3 - 4 229 66
BX933789.2 - 3 - 4 160 66
CR353722.1 - 3 - 4 243 66
CR385458.1 - 2 - 3 340 66
L07034.1 - 2 - 3 154 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

leucine catabolic process
lipid metabolic process
ketone body biosynthetic process
hydroxymethylglutaryl-CoA lyase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0