This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03313273_m1
See other ATP5S GE Assays ›
Gene Symbol:
ATP5S
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases:
-
Chromosome Location:
Chr.5: 57934478 - 57939406 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03313273_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5S
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277562.1 NP_001264491.1 1 - 2 181 144
XM_015276734.2 XP_015132220.1 1 - 2 215 144
GenBank mRNA BU356089.1 - 1 - 2 178 144
BX931650.2 - 1 - 2 181 144
BX932166.2 - 1 - 2 169 144
BX932359.2 - 1 - 2 178 144

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001277562.1, XM_015276734.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0