This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03313347_m1
See other NUDT5 GE Assays ›
Gene Symbol:
NUDT5
Gene Name:
nudix hydrolase 5
Gene Aliases:
YSA1, YSA1H, YSAH1, hYSAH1
Chromosome Location:
Chr.1: 6850849 - 6862491 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03313347_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278105.2 NP_001265034.1 5 - 6 446 121
NM_001278106.1 NP_001265035.1 5 - 6 414 121
NM_001278107.1 NP_001265036.1 4 - 5 320 121
XM_015296341.2 XP_015151827.1 4 - 5 405 121
XM_015296387.2 XP_015151873.1 6 - 7 488 121
XM_015296422.2 XP_015151908.1 5 - 6 455 121
XM_015296473.2 XP_015151959.1 4 - 5 383 121
XM_015296523.2 XP_015152009.1 3 - 4 243 121
XM_025150979.1 XP_025006747.1 5 - 6 416 121
XM_025150989.1 XP_025006757.1 4 - 5 371 121
GenBank mRNA BU250863.1 - 2 - 3 125 121
BU399332.1 - 4 - 5 320 121
BX932306.2 - 3 - 4 248 121
BX932613.2 - 4 - 5 320 121
CR352532.1 - 5 - 6 300 121
CR353333.1 - 4 - 5 224 121
CR353644.1 - 5 - 6 394 121
CR407055.1 - 5 - 6 440 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

pyrophosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0