This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03313608_m1
See other PPP1R7 GE Assays ›
Gene Symbol:
PPP1R7
Gene Name:
protein phosphatase 1 regulatory subunit 7
Gene Aliases:
-
Chromosome Location:
Chr.9: 5097903 - 5112946 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03313608_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277428.1 NP_001264357.1 8 - 9 787 72
NM_001277429.1 NP_001264358.1 7 - 8 709 72
NM_001277430.1 NP_001264359.1 7 - 8 694 72
XM_015276918.2 XP_015132404.1 8 - 9 867 72
XM_015276919.2 XP_015132405.1 8 - 9 769 72
XM_015276920.2 XP_015132406.1 8 - 9 852 72
XM_015276922.2 XP_015132408.1 8 - 9 782 72
XM_025153351.1 XP_025009119.1 8 - 9 759 72
GenBank mRNA BX933967.2 - 4 - 5 324 72
BX934398.2 - 6 - 7 641 72
BX935125.2 - 8 - 9 769 72
BX936156.1 - 7 - 8 683 72
DR413870.1 - 7 - 8 752 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline