This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03313703_m1
See other EEF1E1 GE Assays ›
Gene Symbol:
EEF1E1
Gene Name:
eukaryotic translation elongation factor 1 epsilon 1
Gene Aliases:
EEF1E1-BLOC1S5
Chromosome Location:
Chr.2: 64294340 - 64311277 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03313703_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_418953.6 XP_418953.5 2 - 3 370 88
GenBank mRNA BX934973.2 - 2 - 3 323 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0