This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03314121_m1
See other ATP5C1 GE Assays ›
Gene Symbol:
ATP5C1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
Gene Aliases:
ATP5F1C
Chromosome Location:
Chr.1: 4815817 - 4822605 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03314121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5C1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278096.1 NP_001265025.1 6 - 7 667 137
NM_001278097.1 NP_001265026.1 6 - 7 667 137
XM_015293766.2 XP_015149252.1 6 - 7 707 137
GenBank mRNA BU132888.1 - 6 - 7 632 137
BX936136.1 - 6 - 7 654 137
BX950663.1 - 6 - 7 635 137
CR353583.1 - 6 - 7 667 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0