This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03314674_m1
See other RPS24 GE Assays ›
Gene Symbol:
RPS24
Gene Name:
ribosomal protein S24
Gene Aliases:
-
Chromosome Location:
Chr.6: 14000232 - 14006196 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03314674_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277342.1 NP_001264271.1 6 - 7 484 148
NM_001277343.1 NP_001264272.1 6 - 7 481 148
NM_001277344.1 NP_001264273.1 5 - 6 463 148
GenBank mRNA BU219490.1 - 4 - 5 455 148
BX934192.1 - 5 - 6 455 148
BX934456.2 - 6 - 7 467 148
CR386795.1 - 4 - 5 435 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline