This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03316144_m1
See other NUDT9 GE Assays ›
Gene Symbol:
NUDT9
Gene Name:
nudix hydrolase 9
Gene Aliases:
-
Chromosome Location:
Chr.4: 45696106 - 45724268 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03316144_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004941097.3 XP_004941154.1 7 - 8 897 64
XM_015276509.2 XP_015131995.1 7 - 8 803 64
XM_015276510.2 XP_015131996.1 7 - 8 789 64
XM_025150380.1 XP_025006148.1 8 - 9 994 64
XM_025150381.1 XP_025006149.1 8 - 9 955 64
XM_025150382.1 XP_025006150.1 7 - 8 1247 64
XM_420546.6 XP_420546.4 7 - 8 980 64
GenBank mRNA CR389303.1 - 6 - 7 573 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0