This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03317096_m1
See other BATF GE Assays ›
Gene Symbol:
BATF
Gene Name:
basic leucine zipper ATF-like transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.5: 38492011 - 38497590 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03317096_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_025150893.1 XP_025006661.1 2 - 3 66
GenBank mRNA CR523179.1 - 2 - 3 353 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0