This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03319024_m1
See other POLR2I GE Assays ›
Gene Symbol:
POLR2I
Gene Name:
RNA polymerase II subunit I
Gene Aliases:
-
Chromosome Location:
Chr.16: 1750429 - 1752268 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03319024_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2I
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004951084.2 XP_004951141.1 2 - 3 349 78
XM_423952.6 XP_423952.3 3 - 4 241 78
GenBank mRNA BX933030.2 - 3 - 4 185 78
BX934278.2 - 2 - 3 253 78
CR391498.1 - 3 - 4 184 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0