This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03319850_m1
See other MLLT11 GE Assays ›
Gene Symbol:
MLLT11
Gene Name:
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
Gene Aliases:
-
Chromosome Location:
Chr.25: 230709 - 233283 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03319850_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MLLT11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004948350.3 XP_004948407.1 1 - 2 118 62
GenBank mRNA BX932123.1 - 1 - 2 125 62
BX934266.1 - 1 - 2 103 62
D26324.1 - 1 - 2 105 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_004948350.3 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Gallus gallus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0