This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03320430_m1
See other NAXD GE Assays ›
Gene Symbol:
NAXD
Gene Name:
NAD(P)HX dehydratase
Gene Aliases:
CARKD
Chromosome Location:
Chr.1: 140453421 - 140506478 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03320430_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAXD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004938537.3 XP_004938594.1 2 - 3 304 83
XM_015277872.2 XP_015133358.1 4 - 5 1198 83
XM_015277873.2 XP_015133359.1 4 - 5 1581 83
XM_015277874.1 XP_015133360.1 2 - 3 250 83
GenBank mRNA BX929652.2 - 2 - 3 488 83
BX931345.1 - 2 - 3 170 83
BX934093.1 - 2 - 3 190 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

dehydratase lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0