This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03320871_m1
See other UBE2Q2 GE Assays ›
Gene Symbol:
UBE2Q2
Gene Name:
ubiquitin conjugating enzyme E2 Q2
Gene Aliases:
-
Chromosome Location:
Chr.10: 3950815 - 3989224 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID Gg03320871_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2Q2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015279016.1 XP_015134502.1 11 - 12 1076 68
XM_015279017.2 XP_015134503.1 11 - 12 1312 68
XM_015279018.1 XP_015134504.1 11 - 12 1052 68
XM_015279019.2 XP_015134505.1 11 - 12 1286 68
GenBank mRNA BX931952.1 - 6 - 7 456 68
BX933906.1 - 12 - 13 1004 68
BX935489.1 - 2 - 3 142 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-4gzsq:80/100.66.74.9:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0