This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Gg03322094_s1
See other HDHD3 GE Assays ›
Gene Symbol:
HDHD3 ALAD
Gene Name:
haloacid dehalogenase like hydrolase domain containing 3
aminolevulinate dehydratase
Gene Aliases:
-
ALADH, PBGS
Chromosome Location:
Chr.17: 1819405 - 1820490 on Build GRCg6a
Chromosome Location:
Chr.17: 1812008 - 1819355 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
-
Assay ID Gg03322094_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BX931319.2 - 1 753 73
CR387799.1 - 1 1440 73
Gene Symbol: ALAD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BU394355.1 - 1 528 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline